Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSF2 Antibody (3E2), Alexa Fluor™ 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB11096434AF405
Description
HSF2 Monoclonal antibody specifically detects HSF2 in Human, Mouse, Rat, Porcine, Bovine, Canine, Guinea Pig, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
HSF2 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, Canine, Guinea Pig | |
Purified |
Western Blot, Immunofluorescence | |
300 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
heat shock factor 2, heat shock factor protein 2, heat shock transcription factor 2MGC75048, HSF 2, HSTF 2, HSTF2, MGC117376, MGC156196 | |
Purified recombinant mouse HSF2 protein | |
0.1 mL | |
Cellular Signaling, DNA replication Transcription Translation and Splicing, Epigenetics | |
3298 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction