Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSF2 Antibody (SR2042), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322062100UL
Description
HSF2 Monoclonal antibody specifically detects HSF2 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
HSF2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
SR2042 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
heat shock factor 2, heat shock factor protein 2, heat shock transcription factor 2MGC75048, HSF 2, HSTF 2, HSTF2, MGC117376, MGC156196 | |
A synthesized peptide derived from human HSF2 (Uniprot #: Q03933) | |
100 μg | |
Cellular Signaling, DNA replication Transcription Translation and Splicing, Epigenetics | |
3298 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction