Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSF2 Monoclonal Antibody (3E2)
GREENER_CHOICE

Catalog No. MA191597
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA191597 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA191597 Supplier Invitrogen™ Supplier No. MA191597
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Recommended positive controls: LS174T, SKBR-3, or NIH-3T3 Cells.

Prokaryotic and eukaryotic cells respond to thermal and chemical stress by inducing a group of genes collectively designated heat shock genes. In eukaryotes, this gene expression is regulated primarily at the transcription level. Heat shock transcription factors (HSF, also designated HSTF) 1 and 2 are involved in this regulation. HSF1 and HSF2 are upregulated by estrogen at both the mRNA and protein level. HSF1 is normally found as a monomer whose transcriptional activity is repressed by constitutive phosphorylation. Upon activation, HSF1 forms trimers, gains DNA binding activity and is translocated to the nucleus. HSF2 activity is associated with differentiation and development, and, like HSF1, binds DNA as a trimer. Both HSF1 and HSF2 are known to be induced by proteasome inhibitors of the ubiquitin pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSF2
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3E2
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.1% sodium azide; pH 7.4
Gene HSF2
Gene Accession No. P38533, Q03933
Gene Alias AI661205; heat shock factor 2; heat shock factor protein 2; Heat shock transcription factor 2; HSF 2; Hsf2; HSTF 2; HSTF2; MGC117376; MGC156196; MGC75048
Gene Symbols HSF2
Host Species Rat
Immunogen Recombinant mouse HSF2 protein (aa1-517).
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15500, 3298
Target Species Human, Mouse, Monkey
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.