Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSF2 Monoclonal Antibody (OTI2A7), TrueMAB™, OriGene
SDP

Catalog No. 501681548
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1548 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1548 Supplier OriGene Supplier No. TA807434
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

All organisms respond to elevated temperatures and a variety of environmental stresses by rapid synthesis of heat shock RNAs and proteins. The regulation of heat shock gene transcription is mediated by the transcriptional activator, heat shock factor (HSF), which binds to heat shock response elements (HSEs). Heat shock factor protein 2 (HSF-2) is a DNA-binding protein that specifically binds heat shock promoter elements (HSE) and activates transcription. In higher eukaryotes, HSF is unable to bind to theHSE unless the cells are heat shocked.

Specifications

Antigen HSF2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2A7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene HSF2
Gene Accession No. Q03933
Gene Alias HSF 2, HSTF 2
Gene Symbols HSF2
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 74-324 of human HSF2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3298
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.