Learn More
HSF2 Mouse anti-Human, Clone: OTI4G9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF807406

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
All organisms respond to elevated temperatures and a variety of environmental stresses by rapid synthesis of heat shock RNAs and proteins. The regulation of heat shock gene transcription is mediated by the transcriptional activator, heat shock factor (HSF), which binds to heat shock response elements (HSEs). Heat shock factor protein 2 (HSF-2) is a DNA-binding protein that specifically binds heat shock promoter elements (HSE) and activates transcription. In higher eukaryotes, HSF is unable to bind to theHSE unless the cells are heat shocked.Specifications
HSF2 | |
Monoclonal | |
Unconjugated | |
HSF2 | |
HSF 2, HSTF 2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3298 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4G9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q03933 | |
HSF2 | |
Human recombit protein fragment corresponding to amino acids 74-324 of human HSF2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.