Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSF2 Antibody (OTI3E1), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270948UV
Description
HSF2 Monoclonal specifically detects HSF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
HSF2 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HSF2 | |
Recombinant protein fragment corresponding to amino acids 74-324 of human HSF2 (NP_004497) produced in E.coli. | |
0.1 mL | |
3298.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3E1 | |
Western Blot, Immunohistochemistry | |
heat shock factor 2, heat shock factor protein 2, heat shock transcription factor 2MGC75048, HSF 2, HSTF 2, HSTF2, MGC117376, MGC156196 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction