Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSF2BP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540978
Description
Peptide sequence: RHMGTKEEFV KVRKKDLERL TTEVMQIRDF LPRILNGEVL ESFQKLKIVE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 100%.
HSF2 binding protein (HSF2BP) associates with HSF2. The interaction occurs between the trimerization domain of HSF2 and the amino terminal hydrophilic region of HSF2BP that comprises two leucine zipper motifs. HSF2BP may therefore be involved in modulating HSF2 activation.
Specifications
| HSF2BP | |
| Polyclonal | |
| Unconjugated | |
| HSF2BP | |
| 4932437G14Rik; heat shock factor 2-binding protein; heat shock transcription factor 2 binding protein; Hsf2bp; RGD1565879 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11077 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O75031 | |
| HSF2BP | |
| Synthetic peptide directed towards the N-terminal of human HSF2BP (aa 12-61). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction