Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hsf3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568899
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568899 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568899 Supplier Invitrogen™ Supplier No. PA568899
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Mouse: 100%; Rat: 79%.

Hsf3 gene ontology annotations related to this gene include cellular response to heat; regulation of transcription by RNA polymerase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hsf3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Hsf3
Gene Accession No. D0VYS2
Gene Alias B230358A15Rik; Heat shock factor protein 3; heat shock transcription factor 3; HSF 3; hsf3; HSTF 3; mHSF3
Gene Symbols Hsf3
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse Hsf3 (aa 239-288).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245525
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.