Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSF4 Antibody (2E7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | HSF4 |
---|---|
Clone | 20000000 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
Description
HSF4 Monoclonal specifically detects HSF4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
HSF4 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, CyTOF | |
Unconjugated | |
RUO | |
PBS with 0.05% Sodium Azide | |
HSF4 | |
IgG1 | |
Affinity Purified |
20000000 | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
Monoclonal | |
Mouse | |
Human | |
3299 | |
Purified recombinant fragment of human HSF4 expressed in E. Coli. | |
Primary | |
53 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title