Learn More
Invitrogen™ HSF4 Monoclonal Antibody (2E7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA517090
Description
MA5-17090 targets HSF4 in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17090 immunogen is purified recombinant fragment of human HSF4 expressed in E. Coli. MA5-17090 detects HSF4 which has a predicted molecular weight of approximately 53kDa.
Heat-shock transcription factors (HSFs) activate heat-shock response genes under conditions of heat or other stresses. HSF4 lacks the carboxyl-terminal hydrophobic repeat which is shared among all vertebrate HSFs and has been suggested to be involved in the negative regulation of DNA binding activity. Two alternatively spliced transcripts encoding distinct isoforms and possessing different transcriptional activity have been described.
Specifications
HSF4 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q9ULV5 | |
HSF4 | |
Purified recombinant fragment of human HSF4 expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
2E7 | |
Unconjugated | |
HSF4 | |
ATHSF4; AT-HSFB1; binds heat shock DNA elements; cataract; CTM; CTRCT5; DNA binding protein; heat shock factor 4; heat shock factor protein 4; Heat shock transcription factor 4; hHSF4; HSF 4; Hsf4; HSFB1; HSTF 4; HSTF4; ldis1; Marner; mHSF4 | |
Mouse | |
Protein G | |
RUO | |
3299 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.