Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse HSF4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5695SP
This item is not returnable.
View return policy
Description
HSF4 Monoclonal specifically detects HSF4 in Human, Mouse samples. It is validated for Western Blot.Specifications
HSF4 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
BAA13433 | |
HSF4 | |
E. coli-derived recombinant human HSF4 Ser309-Pro463 Accession # BAA13433 | |
25 μg | |
Primary | |
Detects endogenous human and mouse HSF4 in Western blots. | |
Human, Mouse | |
Purified |
Western Blot | |
504524 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ATHSF4, AT-HSFB1, cataract, Marner, CTM, heat shock factor protein 4, heat shock transcription factor 4CTM, hHSF4, HSF 4, HSFB1, HSTF 4 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3299 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction