Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSF4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $371.50
Specifications
Antigen | HSF4 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145408
|
Invitrogen™
PA5145408 |
20 μL |
Each for $191.80
|
|
|||||
FEPA5145407
|
Invitrogen™
PA5145407 |
200 μL |
Each for $371.50
|
|
|||||
Description
Heat-shock transcription factors (HSFs) activate heat-shock response genes under conditions of heat or other stresses. HSF4 lacks the carboxyl-terminal hydrophobic repeat which is shared among all vertebrate HSFs and has been suggested to be involved in the negative regulation of DNA binding activity. Two alternatively spliced transcripts encoding distinct isoforms and possessing different transcriptional activity have been described.Specifications
HSF4 | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q9R0L1, Q9ULV5 | |
26386, 3299 | |
Recombinant Human HSF4 protein, Val171-Arg284 + SPPATACGCGAGHPGRERELQPRGAQECPTA (Accession #Q9ULV5). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
ATHSF4; AT-HSFB1; binds heat shock DNA elements; cataract; CTM; CTRCT5; DNA binding protein; heat shock factor 4; heat shock factor protein 4; Heat shock transcription factor 4; hHSF4; HSF 4; Hsf4; HSFB1; HSTF 4; HSTF4; ldis1; Marner; mHSF4 | |
HSF4 | |
IgG | |
Affinity Chromatography | |
HSF4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title