Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSF5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564126
Description
Immunogen sequence: LNHNPSPSSV VFVQEGPPFS THQVDANIKC QTSSRENILP SEQMGFLISE MGPASKPSED TGLATPARYR EHRSNSQQGK SPDLH Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 80%.
HSF5 may act as a transcriptional factor.
Specifications
HSF5 | |
Polyclonal | |
Unconjugated | |
Hsf5 | |
Gm739; heat shock factor protein 5; Heat shock transcription factor 5; heat shock transcription factor family member 5; HSF 5; HSF5; HSTF 5; HSTF5; LOW QUALITY PROTEIN: heat shock factor protein 5; MLZ-220; Protein expressed in male leptotene and zygotene spermatocytes 220 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
124535 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q4G112 | |
Hsf5 | |
Recombinant protein corresponding to Human HSF5. Recombinant protein control fragment (Product #RP-103965). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction