Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSFX1 Monoclonal Antibody (1D7)
GREENER_CHOICE

Catalog No. PIMA521924
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521924 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521924 Supplier Invitrogen™ Supplier No. MA521924
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MEDKRSLSMA RCEERNSRGQ DHGLERVPFP PQLQSETYLH PADPSPAWDD PGSTGSPNLR LLTEEIAFQP LAEEASFRRP HPDGDVPPQG EDNLLSLPFP QKLWRLVSSN QFSSIWWDDS GACRVINQKL FEKEILKRDV AHKVFATTSI KSFFRQLNLY GFRKRRQCTF RTFTRIFSAK RLVSILNKLE FYCHPYFQRD SPHLLVRMKR RVGVKSAPRH QEEDKPEAAG SCLAPADTEQ QDHTSPNEND QVTPQHREPA GPNTQIRSGS APPATPVMVP DSAVASDNSP VTQPAGEWSE GSQAHVTPVA AVPGPAALPF LYVPGSPTQM NSYGPVVALP TASRSTLAMD TTGLPAPGML PFCHLWVPVT LVAAGAAQPA ASMVMFPHLP ALHHHCPHSH RTSQYMPASD GPQAYPDYAD QST.

HSFX1 (Heat Shock Transcription Factor Family, X-Linked 1) is a Protein Coding gene. Diseases associated with HSFX1 include Microsporidiosis. Gene Ontology (GO) annotations related to this gene include DNA-binding transcription factor activity and sequence-specific DNA binding. An important paralog of this gene is HSFX2. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSFX1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D7
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene HSFX1
Gene Accession No. Q9UBD0
Gene Alias heat shock transcription factor family, X linked 1; heat shock transcription factor family, X-linked 1; heat shock transcription factor, X-linked; HSFX1; HSFX2; LW-1; ORC4L; testicular secretory protein Li 21
Gene Symbols HSFX1
Host Species Mouse
Immunogen HSFX1 (AAH21706, 1 a.a. ~ 423 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100506164
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.