Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSFX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584581
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584581 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584581 Supplier Invitrogen™ Supplier No. PA584581
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SMARCEERNS RGQDHGLERV PFPPQLQSET YLHPADPSPA WDDPGSTGSP NLRLLTEEIA FQPLAEEASF RRPHPDGDVP PQGEDNLLSL PFPQKLWRLV SSN Highest antigen sequence indentity to the following orthologs: Mouse - 28%, Rat - 28%.

HSFX1 (Heat Shock Transcription Factor Family, X-Linked 1) is a Protein Coding gene. Diseases associated with HSFX1 include Microsporidiosis. Gene Ontology (GO) annotations related to this gene include DNA-binding transcription factor activity and sequence-specific DNA binding. An important paralog of this gene is HSFX2. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSFX1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HSFX1
Gene Accession No. Q9UBD0
Gene Alias heat shock transcription factor family, X linked 1; heat shock transcription factor family, X-linked 1; heat shock transcription factor, X-linked; HSFX1; HSFX2; LW-1; ORC4L; testicular secretory protein Li 21
Gene Symbols HSFX1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HSFX1. Recombinant protein control fragment (Product #RP-108482).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100506164
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.