Learn More
Invitrogen™ HSFY1/HSFY2 Polyclonal Antibody

Description
Recommended positive controls: mouse testis, Rat testis. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | HSFY1/HSFY2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | HSFY1 |
| Gene Accession No. | Q96LI6 |
| Gene Alias | 4933413G11Rik; Heat shock transcription factor 2-like protein; heat shock transcription factor, Y linked 2; heat shock transcription factor, Y linked 2 like; heat shock transcription factor, Y-linked; heat shock transcription factor, Y-linked 1; heat shock transcription factor, Y-linked 2; HSF2L; HSF2-like; HSFY; Hsfy1; HSFY2; Hspy2l |
| Gene Symbols | HSFY1, Hsfy2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.