Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSFY1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541190 Supplier Invitrogen™ Supplier No. PA541190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SWDENGTCIV INEELFKKEI LETKAPYRIF QTDAIKSFVR QLNLYGFSKI Sequence homology: Cow: 79%; Dog: 79%; Human: 100%; Mouse: 79%; Rat: 79%.

This gene encodes a member of the heat shock factor (HSF) family of transcriptional activators for heat shock proteins. This gene is a candidate gene for azoospermia, since it localizes to a region of chromosome Y that is sometimes deleted in infertile males. The genome has two identical copies of this gene within a palindromic region; this record represents the more centromeric copy. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSFY1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HSFY1
Gene Accession No. Q96LI6
Gene Alias Heat shock transcription factor 2-like protein; heat shock transcription factor, Y-linked; heat shock transcription factor, Y-linked 1; HSF2L; HSF2-like; HSFY; HSFY1; HSFY2; Y-linked 1
Gene Symbols HSFY1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human HSFY1 (aa 97-146).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 86614
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.