Learn More
HSG, Heat sink grease
Description
Thermally conductive heat sink grease applied to the base and mounting stud of transistors, diodes and rectifiers, as a coupling agent, to reduce thermal contact resistance. Ideal for sputtering target applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Density | 2.3 |
| Odor | Odorless |
| Quantity | 16 oz. |
| Physical Form | Paste |
| Chemical Name or Material | HSG |
Safety and Handling
| Hazard Category | H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.