Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSH2D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578608
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578608 Supplier Invitrogen™ Supplier No. PA578608
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: NCI-H929 Predicted Reactivity: Rhesus Monkey (96%), Chimpanzee (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

T-cell activation requires 2 signals: recognition of antigen by the T-cell receptor and a costimulatory signal provided primarily by CD28 in naive T cells. HSH2 is a target of both of these signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSH2D
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene HSH2D
Gene Accession No. Q96JZ2
Gene Alias Adaptor in lymphocytes of unknown function X; ALX; FLJ14886; hematopoietic SH2 domain containing; hematopoietic SH2 domain-containing protein; Hematopoietic SH2 protein; HSH2; HSH2D
Gene Symbols HSH2D
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human HSH2D. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84941
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.