Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSJ1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987764
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7764 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7764 Supplier Bioss Supplier No. BS7233R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The DnaJ family is one of the largest of all the chaperone families and has evolved with diverse cellular localization and functions. The presence of the J domain defines a protein as a member of the DnaJ family. DnaJ heat shock induced proteins are from the bacterium Escherichia coli and are under the control of the htpR regulatory protein. The DnaJ proteins play a critical role in the HSP 70 chaperone machine by interacting with HSP 70 to stimulate ATP hydrolysis. The proteins contain cysteine rich regions that are composed of zinc fingers that form a peptide binding domain responsible for the chaperone function. DnaJ proteins are important mediators of proteolysis and are involved in the regulation of protein degradation, exocytosis and endocytosis. DnaJB2 (DnaJ homolog subfamily B member 2), also known as HSJ1 or HSPF3, is expressed almost exclusively in the brain, with the highest levels in the frontal cortex and hippocampus. Two isoforms are produced due to alternative splicing.

Specifications

Antigen HSJ1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DNAJB2
Gene Accession No. P25686
Gene Alias 2700059H22Rik; CMT2T; DnaJ (Hsp40) homolog, subfamily B, member 10; DnaJ (Hsp40) homolog, subfamily B, member 2; DnaJ heat shock protein family (Hsp40) member B2; DnaJ homolog subfamily B member 10; DnaJ homolog subfamily B member 2; dnaJ protein homolog 1; Dnajb10; Dnajb2; DSMA5; Heat shock 40 kDa protein 3; Heat shock protein J1; heat shock protein, DNAJ-like 1; heat shock protein, neuronal DNAJ-like 1; HSJ1; HSJ-1; HSPF3; mDj8
Gene Symbols DNAJB2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HSJ1/DNAJB2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3300
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.