Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115384
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115384 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115384 Supplier Invitrogen™ Supplier No. PA5115384
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HSL.

Hormone-sensitive lipase (HSL) is an intracellular neutral lipase found in a variety of tissues, but primarily in adipose and steroidogenic tissues. It hydrolyzes triglycerides and cholesteryl esters. It is the major enzyme responsible for the mobilization of free fatty acids from adopise tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSL
Applications Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene LIPE
Gene Accession No. P15304, P54310, Q05469
Gene Alias 4933403G17Rik; AOMS4; EC 3.1.1.79; FPLD6; hormone - sensitive lipase testicular isoform; hormone-sensitive lipase; hormone-sensitive lipase testicular isoform; HSL; HSL antibody; LHS; lipase E; lipase E, hormone sensitive type; lipase, hormone sensitive; lipase, hormone-sensitive; LIPE; LIPS
Gene Symbols LIPE
Host Species Rabbit
Immunogen A synthesized peptide derived from human LIPE(Accession Q05469), corresponding to amino acid residues W827-R877.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16890, 25330, 3991
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.