Learn More
Invitrogen™ HSL Polyclonal Antibody

Description
PA1-16966 is specific for the ~84 kDa Hormone-Sensitive Lipase protein. A non-specific band may also be detected at ~89 kDa. Bands at ~102, 113, and 127 kDa may also be seen in the testis of sexually immature rats.Suggested positive control: human adipose protein.
Specifications
Specifications
| Antigen | HSL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | LIPE |
| Gene Accession No. | P15304, P54310, Q05469 |
| Gene Alias | 4933403G17Rik; AOMS4; EC 3.1.1.79; FPLD6; hormone - sensitive lipase testicular isoform; hormone-sensitive lipase; hormone-sensitive lipase testicular isoform; HSL; HSL antibody; LHS; lipase E; lipase E, hormone sensitive type; lipase, hormone sensitive; lipase, hormone-sensitive; LIPE; LIPS |
| Gene Symbols | LIPE |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.