Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP10 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987676
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7676 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7676 Supplier Bioss Supplier No. BS7026R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The heat shock proteins (HSPs) comprise a group of highly conserved, abundantly expressed proteins with diverse functions, including the assembly and sequestering of multiprotein complexes, transportation of nascent polypeptide chains across cellular membranes and regulation of protein folding. Heat shock proteins (also known as molecular chaperones) fall into six general families: HSP 90, HSP 70, HSP 60, the low molecular weight HSPs, the immunophilins and the HSP 110 family. The low molecular weight family includes HSP 10, HSP 20, HSP 27 (Heme Oxygenase 1), HSP 32 and HSP 40. Chaperonins are ubiquitous, indispensable proteins that facilitate protein folding in an ATP-dependent manner, enhancing the yield of properly folded substrate protein under conditions where spontaneous folding does not occur. Chaperonins are typified by the E. coli heat-shock proteins GroEL and GroES (HSP 10). HSP 10 is a heptameric ring of identical 10 kDa subunits that binds to each end of GroEL to form a symmetric, functional heterodimer.

Specifications

Antigen HSP10
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HSPE1
Gene Accession No. P61604
Gene Alias 10 kDa chaperonin; 10 kDa heat shock protein, mitochondrial; 10kDa; 10kDa Chaperonin; BOS_1951; CH10; Chaperonin 10; Cpn 10; CPN10; early pregnancy factor; Early-pregnancy factor; EPF; GROES; Heat shock 10 kD protein 1 (chaperonin 10); heat shock 10 kDa protein 1 (chaperonin 10); heat shock 10 protein 1; heat shock 105kDa/110kDa protein 1; heat shock 10kD protein; heat shock 10kD protein 1 (chaperonin 10); heat shock 10kDa protein 1; heat shock 10kDa protein 1 (chaperonin 10); heat shock protein; heat shock protein 1 (chaperonin 10); heat shock protein 105; heat shock protein 105 kDa; heat shock protein family E (Hsp10) member 1; heat shock protein family E (Hsp10) member 1 L homeolog; heat shock protein family H (Hsp110) member 1; HSP; Hsp10; hsp105; hsp105a; hsp105b; hsp110; Hspe1; hspe1.L; hsph1; hsph1-a; hsph1-b; LOC100350401; LOC100357698; mitochondrial 10 kDa heat shock protein; mitochondrial chaperonin 10; MOB family member 4, phocein; MOB1, Mps One Binder kinase activator-like 3; MOB4; MOBKL3; mob-like protein phocein; mps one binder kinase activator-like 3-like; mt-cpn10; PREI3; preimplantation protein 3; XELAEV_18045050mg; zgc:86747
Gene Symbols HSPE1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HSP10.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3336
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.