Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSP104 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA1024

Catalog No. PA1-024


Only null left
Edge
Add to Cart

Description

Description

PA1-024 detects HSP104 from yeast samples. PA1-024 has been successfully used in Western blot procedures. By Western blot, this antibody detects a single ~104 kDa protein from yeast. The PA1-024 immunogen is a synthetic peptide corresponding to the C-terminal residues D(894) D D N E D S M E I D D D L D(908) of S. cerevisiae HSP104.

Heat shock proteins (HSP) are expressed in response to various biological stresses, including high temperatures. There are several major families of HSPs including HSP70, HSP90 and HSP100. The HSP100 proteins generally have amino acid sequences of about 900 residues and contain two nucleotide-binding sites. Within the HSP100 family of proteins, yeast express an ~104 kDa form which is necessary to protect cells from various stress conditions such as heat, heavy metals and ethanol, though mutation studies have shown that the protein is not required for normal growth. Yeast HSP104 has been shown to be a ClpB protein with significant sequence homology to E. coli ClpB, particularly in the two nucleotide-binding sites.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HSP104
Polyclonal
Unconjugated
HSP104
chaperone ATPase HSP104; Heat shock protein; Heat shock protein 104; HS104; HSP; HSP104; L0948; Protein aggregation-remodeling factor HSP104; YLL026W
Rabbit
Antigen Affinity Chromatography
RUO
850633
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
P31539
HSP104
Synthetic peptide corresponding to the C-terminal residues D(894) D D N E D S M E I D D D L D(908) of S. cerevisiae HSP104.
100 μg
Primary
Yeast
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.