Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSP20 Monoclonal Antibody (C1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA543766
This item is not returnable.
View return policy
Description
HSP20 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
HSPB6 is associated with actin and modulates smooth muscle relaxation. This locus encodes a heat shock protein. The encoded protein likely plays a role in smooth muscle relaxation.
Specifications
HSP20 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P97541 | |
HSPB6 | |
Recombinant protein Heat Shock Protein Beta 6. The antigen corresponds to amino acid range 1-162 of the target protein. | |
20 μL | |
Primary | |
Rat | |
Antibody | |
IgG2a κ |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
C1 | |
Unconjugated | |
HSPB6 | |
AA387366; cardiac heat shock protein p20; cardiac p20; epididymis luminal protein 55; Gm479; Heat shock 20 kDa-like protein p20; heat shock 20-kDa protein; Heat shock protein; heat shock protein 20kDa; heat shock protein beta-6; heat shock protein family B (small) member 6; heat shock protein family B (small) member B6; heat shock protein, alpha-crystallin-related, B6; HEL55; HSP; HSP20; HSPB6; p20; PPP1R91; protein phosphatase 1, regulatory subunit 91 | |
Mouse | |
Protein A/G | |
RUO | |
192245 | |
-20°C | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction