Learn More
Invitrogen™ HSP20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595454
Description
Human Hsp20 shares 88.2% and 92.2% amino acid (aa) sequence identity with mouse and rat Hsp20, respectively. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat cardiac muscle tissue, rat skeletal muscle tissue, rat testis tissue, mouse testis tissue, human A431 whole cell. IHC: human skeletal muscle tissue, rat cardiac muscle tissue, human mammary cancer tissue, mouse lung tissue, mouse cardiac muscle tissue, rat lung tissue. ICC/IF: A431 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
HSPB6 is associated with actin and modulates smooth muscle relaxation. This locus encodes a heat shock protein. The encoded protein likely plays a role in smooth muscle relaxation.
Specifications
HSP20 | |
Polyclonal | |
Unconjugated | |
HSPB6 | |
AA387366; cardiac heat shock protein p20; cardiac p20; epididymis luminal protein 55; Gm479; Heat shock 20 kDa-like protein p20; heat shock 20-kDa protein; Heat shock protein; heat shock protein 20kDa; heat shock protein beta-6; heat shock protein family B (small) member 6; heat shock protein family B (small) member B6; heat shock protein, alpha-crystallin-related, B6; HEL55; HSP; HSP20; HSPB6; p20; PPP1R91; protein phosphatase 1, regulatory subunit 91 | |
Rabbit | |
Affinity chromatography | |
RUO | |
126393, 192245, 243912 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
O14558, P97541, Q5EBG6 | |
HSPB6 | |
E.coli-derived human Hsp20 recombinant protein (Position: H73-L123). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.