Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP20 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984034
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4034 Supplier Bioss Supplier No. BS1688R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HSPB6 is associated with actin and modulates smooth muscle relaxation. This locus encodes a heat shock protein. The encoded protein likely plays a role in smooth muscle relaxation.

Specifications

Antigen HSP20
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HSPB6
Gene Accession No. O14558
Gene Alias AA387366; BOS_17547; cardiac heat shock protein p20; cardiac p20; epididymis luminal protein 55; Gm479; Heat shock 20 kDa-like protein p20; heat shock 20-kDa protein; Heat shock protein; heat shock protein 20kDa; Heat shock protein beta-6; heat shock protein family B (small) member 6; heat shock protein family B (small) member B6; heat shock protein, alpha-crystallin-related, B6; HEL55; HSP; HSP20; Hspb6; LOC100347944; LOC494560; p20; PPP1R91; protein phosphatase 1, regulatory subunit 91
Gene Symbols HSPB6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HSP20.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126393
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.