Learn More
Invitrogen™ HSP20 Recombinant Rabbit Monoclonal Antibody (3J5N1)

Description
Positive Samples: Mouse heart, Rat skeletal muscle Immunogen sequence: DPGHFSVLLD VKHFSPEEIA VKVVGEHVEV HARHEERPDE HGFVAREFHR RYRLPPGVDP AAVTSALSPE GVLSIQAAPA S.
Specifications
Specifications
| Antigen | HSP20 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 3J5N1 |
| Concentration | 0.34 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | HSPB6 |
| Gene Accession No. | O14558, P97541, Q5EBG6 |
| Gene Alias | AA387366; cardiac heat shock protein p20; cardiac p20; epididymis luminal protein 55; Gm479; Heat shock 20 kDa-like protein p20; heat shock 20-kDa protein; Heat shock protein; heat shock protein 20kDa; heat shock protein beta-6; heat shock protein family B (small) member 6; heat shock protein family B (small) member B6; heat shock protein, alpha-crystallin-related, B6; HEL55; HSP; HSP20; HSPB6; p20; PPP1R91; protein phosphatase 1, regulatory subunit 91 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.