Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP27 Antibody (6H11), Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $499.50
Specifications
Antigen | HSP27 |
---|---|
Clone | 6H11 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:10000, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, CyTOF-ready, Knockout Validated |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF, KnockDown |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP225149SS
![]() |
Novus Biologicals
NBP225149SS |
0.025 mL |
Each for $243.50
|
|
|||||
NBP225149
![]() |
Novus Biologicals
NBP225149 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
HSP27 Monoclonal specifically detects HSP27 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, CyTOF-ready, Knockout Validated.Specifications
HSP27 | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF, KnockDown | |
Unconjugated | |
RUO | |
Human, Mouse (Negative), Rat (Negative) | |
P04792 | |
3315 | |
Recombinant full length human HSP27 expressed in and purified from E. coli. [UniProt# P04792] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
27 kDa |
6H11 | |
Western Blot 1:10000, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, CyTOF-ready, Knockout Validated | |
Monoclonal | |
Mouse | |
Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific | |
50% PBS, 50% glycerol with 5mM Sodium Azide | |
28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27 | |
HSPB1 | |
IgG1 | |
Affinity Purified | |
Human, Bovine, Porcine, Mouse and Rat. Other species have not been tested. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title