Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP27 Antibody (6H11), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP225149PCP
Description
HSP27 Monoclonal specifically detects HSP27 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Knockout Validated.Specifications
HSP27 | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
HSPB1 | |
Recombinant full length human HSP27 expressed in and purified from E. coli. [UniProt# P04792] | |
0.1 mL | |
Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific | |
3315 | |
Store at 4C in the dark. |
Flow Cytometry, Immunoassay, Immunocytochemistry, Immunofluorescence | |
6H11 | |
Flow Cytometry, Knockout Validated | |
28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse (Negative), Rat (Negative) | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction