Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP27 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP175477AF594
Description
HSP27 Polyclonal specifically detects HSP27 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
HSP27 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated | |
28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27 | |
Rabbit | |
Affinity Purified | |
Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific | |
3315.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
HSPB1 | |
A synthetic peptide made to an internal region of the human Hsp27 protein (within residues 80-150). [Swiss-ProtNo. P04792] | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction