Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSP27 Monoclonal Antibody (6H11)

Catalog No. PIMA547365
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547365 500 μL
PIMA547464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA547365 Supplier Invitrogen™ Supplier No. MA547365
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HSP27 Monoclonal Antibody for Western Blot, ICC/IF, IHC

In response to adverse changes in their environment, cells from many organisms increase the expression of a class of proteins referred to as heat shock or stress proteins. HSPB1 (heat shock protein beta-1 or HSP27) is a small heat shock protein which functions as a molecular chaperone that maintains denatured proteins in a folding-competent state. It plays a role in stress resistance and actin organization. Through its molecular chaperone activity, HSP27 regulates numerous biological processes including the phosphorylation and the axonal transport of neurofilament proteins. Mutations in the gene can result in Charcot-Marie-Tooth disease 2F and Neuronopathy distal hereditary motor 2B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP27
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6H11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 5mM sodium azide
Gene HSPB1
Gene Accession No. P04792
Gene Alias 27kDa; 28 kDa heat shock protein; CMT2F; DKFZp586P1322; epididymis secretory protein Li 102; estrogen-regulated 24 kDa protein; Growth-related 25 kDa protein; heat shock 25 kDa protein; Heat shock 27 kDa protein; heat shock 27kD protein 1; heat shock 27kDa protein 1; Heat shock protein; heat shock protein 1; heat shock protein 27 kDa beta-1; heat shock protein B1; heat shock protein beta-1; heat shock protein family B (small) member 1; heat shock protein, 25 kDa; heat-shock protein; HEL-S-102; HMN2B; HS; HS.76067; HSP; HSP 25; HSP 27; Hsp25; HSP27; HSP28; HSPB1; p25; Phospho-HSP 27; SRP27; Stress-responsive protein 27; truncated hsp25
Gene Symbols HSPB1
Host Species Mouse
Immunogen Recombinant full length human HSP27 expressed in and purified from E. coli.
Purification Method Protein A/G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3315
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.