Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSP27 Monoclonal Antibody (STRSN), eBioscience™

Catalog No. 14911280
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-911-280 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-911-280 Supplier Invitrogen™ Supplier No. 14911280
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The monoclonal antibody STRSN recognizes human heat shock protein 27 (HSP27). The heat shock proteins are a large family of proteins that are induced in response to stressors such as changing temperatures, cytokine levels, and presence of various chemicals. Heat shock proteins serve as molecular chaperones, binding to other proteins to maintain proper protein structure or by folding denatured proteins and preventing activation of caspases that could lead to apoptosis. Unlike the larger HSP proteins, HSP27 belongs to a unique subgroup that is involved in apoptosis regulation. Expression of HSP27 is constitutive, but upon stress HSP27 is upregulated and can relocate from the cytoplasm to the nucleus. Increased HSP27 expression with certain carcinomas has been observed. In breast cancer, HSP27 expression has been shown to be responsive to estrogen levels. In both breast cancer and melanoma, high levels of HSP27 correlate with more aggressive tumors and decreased survival. Increased levels of HSP27 in the serum of patients with Chronic Obstructive Pulmonary Disease (COPD), breast cancer and melanoma may serve as a potential diagnostic marker. No crossreactivity is observed with HSP70, 90, 104 or 110. Applications Reported: This STRSN antibody has been reported for use in western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, ELISA, and immunocytochemistry. Applications Tested: This STRSN antibody has been tested by west...

In response to adverse changes in their environment, cells from many organisms increase the expression of a class of proteins referred to as heat shock or stress proteins. HSPB1 (heat shock protein beta-1 or HSP27) is a small heat shock protein which functions as a molecular chaperone that maintains denatured proteins in a folding-competent state. It plays a role in stress resistance and actin organization. Through its molecular chaperone activity, HSP27 regulates numerous biological processes including the phosphorylation and the axonal transport of neurofilament proteins. Mutations in the gene can result in Charcot-Marie-Tooth disease 2F and Neuronopathy distal hereditary motor 2B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP27
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone STRSN
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene HSPB1
Gene Accession No. P04792
Gene Alias 27kDa; 28 kDa heat shock protein; CMT2F; DKFZp586P1322; epididymis secretory protein Li 102; estrogen-regulated 24 kDa protein; Growth-related 25 kDa protein; heat shock 25 kDa protein; Heat shock 27 kDa protein; heat shock 27kD protein 1; heat shock 27kDa protein 1; Heat shock protein; heat shock protein 1; heat shock protein 27 kDa beta-1; heat shock protein B1; heat shock protein beta-1; heat shock protein family B (small) member 1; heat shock protein, 25 kDa; heat-shock protein; HEL-S-102; HMN2B; HS; HS.76067; HSP; HSP 25; HSP 27; Hsp25; HSP27; HSP28; HSPB1; p25; Phospho-HSP 27; SRP27; Stress-responsive protein 27; truncated hsp25
Gene Symbols HSPB1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3315
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.