Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP27 Antibody (017), Biotin

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289442B
Description
HSP27 Monoclonal specifically detects HSP27 in Human samples. It is validated for Western Blot, ELISA.Specifications
HSP27 | |
Monoclonal | |
Biotin | |
28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human HSP27 (NP_001532.1; Met 1-Lys 205) . | |
0.1 mL | |
Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific | |
3315 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
17 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction