Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP40/DNAJB1 Antibody (2A7.H6), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP122416AF488
Description
HSP40/DNAJB1 Monoclonal specifically detects HSP40/DNAJB1 in Yeast samples. It is validated for Western Blot, ELISA.Specifications
HSP40/DNAJB1 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DnaJ (Hsp40) homolog, subfamily B, member 1, DnaJ (Hsp40) homolog, subfmaily B, member 1, dnaJ homolog subfamily B member 1, DnaJ protein homolog 1, DNAJ1, hDj-1, HDJ1, Heat shock 40 kDa protein 1, heat shock 40kD protein 1, Heat shock protein 40, HSP40, HSPF1Hdj1, Human DnaJ protein 1, radial spoke 16 homolog B, RSPH16B, Sis1 | |
Mouse | |
Protein G purified | |
DNA Repair, Golgi Apparatus Markers, Membrane Trafficking and Chaperones | |
3337 | |
Yeast | |
Purified |
Western Blot, ELISA | |
2A7.H6 | |
Western Blot, ELISA | |
P25491 | |
DNAJB1 | |
Full length protein HSP40 (YDJ1) | |
0.1 mL | |
Primary | |
Detects approx 40kDa. Yeast specific. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction