Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP40/DNAJB1 Antibody (2A7.H6), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP122416
Description
HSP40/DNAJB1 Monoclonal specifically detects HSP40/DNAJB1 in Yeast samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
HSP40/DNAJB1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
DnaJ (Hsp40) homolog, subfamily B, member 1, DnaJ (Hsp40) homolog, subfmaily B, member 1, dnaJ homolog subfamily B member 1, DnaJ protein homolog 1, DNAJ1, hDj-1, HDJ1, Heat shock 40 kDa protein 1, heat shock 40kD protein 1, Heat shock protein 40, HSP40, HSPF1Hdj1, Human DnaJ protein 1, radial spoke 16 homolog B, RSPH16B, Sis1 | |
Mouse | |
Protein G purified | |
RUO | |
3337 | |
Yeast | |
Purified |
Western Blot, ELISA, Immunoprecipitation | |
2A7.H6 | |
Unconjugated | |
P25491 | |
DNAJB1 | |
Full length protein HSP40 (YDJ1) | |
0.1 mg | |
Primary | |
Detects approx 40kDa. Yeast specific. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction