Learn More
Invitrogen™ HSP40 Monoclonal Antibody (1G10.H8), FITC

Description
0.5 μg/mL of MA5-45187 was sufficient for detection of 50 ng YDJ1 by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 40kDa. Yeast specific product. Does not cross react with Human, Mouse or Rat.
Specifications
Specifications
| Antigen | HSP40 |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 1G10.H8 |
| Concentration | 1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative |
| Gene Alias | 0610007I11Rik; DjB1; DnaJ (Hsp40) homolog, subfamily B, member 1; DnaJ heat shock protein family (Hsp40) member B1; dnaJ homolog subfamily B member 1; DnaJ protein homolog 1; DNAJ1; Dnajb1; HDJ1; hDj-1; Heat shock 40 kDa protein 1; Heat shock protein; HSP; HSP 40; HSP40; HSPF1; human DnaJ protein 1; NDAJB1; radial spoke 16 homolog B; RSPH16B; Sis1 |
| Host Species | Mouse |
| Immunogen | Full length protein yeast HSP40 (YDJ1). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.