Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSP40 Monoclonal Antibody (2A7.H6), FITC
GREENER_CHOICE

Catalog No. PIMA545221
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545221 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545221 Supplier Invitrogen™ Supplier No. MA545221
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

0.5 μg/mL of MA5-45221 was sufficient for detection of 50 ng YDJ1 by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 40kDa. Yeast specific. Does not cross react with Human, Mouse or Rat.

TCF-9 (transcription factor 9), also known as C2orf3 (chromosome 2 open reading frame 3), GCF (GC-rich sequence DNA-binding factor) or DNABF, is a 781 amino acid nuclear protein that belongs to the GCF family. Widely expressed, GCF binds the GC-rich sequences of beta-Actin, EGFR and calcium-dependent protease (CANP) promoters. GCF contains multiple phosphoserine and phosphothreonine residues, and two GCF isoforms are produced due to alternative splicing events. GCF is considered a candidate for susceptibility to dyslexia (DYX3) as both genes reside in close proximity on human chromosome 2. Chromosome 2 is the second largest human chromosome and consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP40
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2A7.H6
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative
Gene Alias 0610007I11Rik; DjB1; DnaJ (Hsp40) homolog, subfamily B, member 1; DnaJ heat shock protein family (Hsp40) member B1; dnaJ homolog subfamily B member 1; DnaJ protein homolog 1; DNAJ1; Dnajb1; HDJ1; hDj-1; Heat shock 40 kDa protein 1; Heat shock protein; HSP; HSP 40; HSP40; HSPF1; human DnaJ protein 1; NDAJB1; radial spoke 16 homolog B; RSPH16B; Sis1
Host Species Mouse
Immunogen Full length protein HSP40 (YDJ1).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.