Learn More
Hsp40/YDJ1 Antibody (1G10.H8), Alexa Fluor™ 700, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP212879AF700
Catalog No. NB212879700
Only null left
Add to Cart
Description
Description
Hsp40/YDJ1 Monoclonal specifically detects Hsp40/YDJ1 in Yeast samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
Hsp40/YDJ1
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
YDJ1
Full length protein yeast HSP40 (YDJ1)
100 μL
855661
Yeast
Purified
Western Blot, ELISA
1G10.H8
Western Blot, ELISA
MAS5, YDJ1 Ydj1p
Mouse
Protein G purified
Primary
Detects approx 40kDa. Yeast specific product.
Store at 4C in the dark.
IgG1 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.