Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hsp47 Antibody (1C4-1A6), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2224000.05MG
Description
Hsp47 Monoclonal specifically detects Hsp47 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Hsp47 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence | |
(collagen binding protein 1), Arsenic-transactivated protein 3, AsTP3, CBP1, CBP2RA-A47, Cell proliferation-inducing gene 14 protein, Collagen-binding protein, colligen, Colligin, colligin-1, colligin-2, gp46, HSP47serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1), member 2, member 2, (collagen-binding protein 2), PPROM, proliferation-inducing gene 14, rheumatoid arthritis antigen A-47,47 kDa heat shock protein, Rheumatoid arthritis-related antigen RA-A47, serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), serpin H1, serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, SERPINH2colligin | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Detects 47kDa. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1C4-1A6 | |
Unconjugated | |
P50454.2 | |
SERPINH1 | |
Human HSP47, full length | |
0.05 mg | |
Core ESC Like Genes, Stem Cell Markers | |
871 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction