Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP60 Antibody (4B9/89) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | HSP60 |
---|---|
Clone | 4B9/89 |
Dilution | Western Blot 1:100 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2 μg, Immunohistochemistry-Paraffin 1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
HSP60 Monoclonal specifically detects HSP60 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
HSP60 | |
Western Blot 1:100 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2 μg, Immunohistochemistry-Paraffin 1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Primate | |
60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant) | |
HSPD1 | |
IgG2a | |
Protein A purified | |
Store at -20°C. Avoid freeze-thaw cycles. |
4B9/89 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Apoptosis, Cellular Markers, Membrane Trafficking and Chaperones, Mitochondrial Markers | |
P10809 | |
3329 | |
Human placental HSP60. | |
Primary | |
Reconstitute with PBS. | |
This antibody reacts with Human, Rat and Primate. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title