Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP60 Antibody (GROEL/730), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234625APC
Description
HSP60 Monoclonal specifically detects HSP60 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry.Specifications
| HSP60 | |
| Monoclonal | |
| APC | |
| 60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant) | |
| Mouse | |
| 60 kDa | |
| 0.1 mL | |
| Apoptosis, Cellular Markers, Membrane Trafficking and Chaperones, Mitochondrial Markers | |
| 3329 | |
| Human, Mouse (Negative), Rat (Negative) | |
| Purified |
| Flow Cytometry | |
| GROEL/730 | |
| Flow Cytometry | |
| HSPD1 | |
| Recombinant human HSP60 protein (Uniprot: P10809) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes a 60kDa protein, identified as the heat shock protein 60 (HSP60). A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of a family of proteins known as heat shock proteins (hsps). Hsp60 is a potential antigen in a number of autoimmune diseases. In human arthritis and in experimentally induced arthritis in animals, disease development coincides with the development of immune reactivity directed against not only bacterial HSP60, but also against its mammalian homolog. Clone GROEL730 reacts only with human and is useful in distinguishing human HSP60 from other mammalian and bacterial HSP60. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction