Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP60 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177397H
Description
HSP60 Polyclonal specifically detects HSP60 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting, Knockdown Validated.Specifications
HSP60 | |
Polyclonal | |
Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting, Knockdown Validated | |
P10809 | |
HSPD1 | |
A synthetic peptide made to an internal region of the human Hsp60 protein (within residues 70-150). [Swiss-Prot P10809] | |
0.1 mL | |
Apoptosis, Cellular Markers, Membrane Trafficking and Chaperones, Mitochondrial Markers | |
3329 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoblot, KnockDown | |
HRP | |
PBS with No Preservative | |
60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant) | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, mouse, rat, hamster and primate. Immunogen has 100% identity to bovine and chicken. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction