Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP60 Antibody (HSPD1/875) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$646.00

Specifications

Antigen HSP60
Clone HSPD1/875
Concentration 1.0 mg/mL
Dilution Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready
Applications Flow Cytometry, Immunofluorescence, CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP247844A
SDP
View Documents
Novus Biologicals
NBP2478440.1MG
0.1 mg
Each for $646.00
Only null left
Add to Cart
 
NBP247844B
SDP
View Documents
Novus Biologicals
NBP2478440.2MG
0.2 mg N/A N/A N/A
Description

Description

HSP60 Monoclonal specifically detects HSP60 in Human, Mouse, Rat, Chicken, Guinea Pig, Hamster, Monkey samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

HSP60
1.0 mg/mL
Flow Cytometry, Immunofluorescence, CyTOF
Unconjugated
Mouse
Apoptosis, Cellular Markers, Membrane Trafficking and Chaperones, Mitochondrial Markers
PBS with No Preservative
3329
Recombinant human HSPD1 protein
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
60 kDa
HSPD1/875
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready
Monoclonal
Purified
RUO
Human, Mouse, Rat, Chicken, Guinea Pig, Hamster, Monkey
60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant)
HSPD1
IgG1 κ
Protein A or G purified
Recognizes a 60kDa protein, identified as the heat shock protein 60 (hsp60). A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of a family of proteins known as heat shock proteins (hsps), more appropriately called as stress response proteins (srps). hsp60 is a potential antigen in a number of autoimmune diseases. In human arthritis and in experimentally induced arthritis in animals, disease development coincides with the development of immune reactivity directed against not only bacterial hsp60, but also against its mammalian homolog.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.