Learn More
Invitrogen™ HSP60 Monoclonal Antibody (A57-E4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA3022
Description
MA3-022 detects heat shock protein 60 kDa (HSP60) from bacteria. This antibody detects HSP60 from all three species of Chlamydia, Escherichia coli (GroEL), Salmonella typhimurium, Neisseria gonorrhoeae and Coxiella burnetti. MA3-022 does not cross react with HeLa cell HSP60. MA3-022 has been successfully used in Western blot procedures. By Western blot, this antibody detects a single ~60 kDa protein representing HSP60 from C. trachomatis lysate. The MA3-022 immunogen is recombinant Chlamydia trachomatis HSP60. Epitope mapping studies suggest that this antibody binds to a linear sequence of 6 amino acids of Chlamydia HSP60 amino acids 117-122.
This gene encodes a member of the chaperonin family. The encoded mitochondrial protein may function as a signaling molecule in the innate immune system. This protein is essential for the folding and assembly of newly imported proteins in the mitochondria. This gene is adjacent to a related family member and the region between the 2 genes functions as a bidirectional promoter. Several pseudogenes have been associated with this gene. Two transcript variants encoding the same protein have been identified for this gene. Mutations associated with this gene cause autosomal recessive spastic paraplegia 13.
Specifications
HSP60 | |
Monoclonal | |
Conc. Not Determined | |
Ascites with 0.05% sodium azide | |
Mouse | |
100 μL | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
A57-E4 | |
Unconjugated | |
60 kDa chaperonin; 60 kDa heat shock protein; 60 kDa heat shock protein, mitochondrial; 60kDa; Chaperonin 60; CPN60; GROEL; heat shock 60kD protein 1 (chaperonin); heat shock 60kDa protein 1; heat shock 60kDa protein 1 (chaperonin); Heat shock protein; heat shock protein 1 (chaperonin); heat shock protein 60; heat shock protein 60 (liver); heat shock protein 65; heat shock protein family D (Hsp60) member 1; heat shock protein, 60 kDa; HLD4; HSP; Hsp60; HSP-60; Hsp60s1; HSP65; HSP-65; Hspd1; Hspd1-30p; HuCHA60; hypothetical protein; I79_011398; mitochondrial matrix protein P1; P1 protein; P60 lymphocyte protein; RCJMB04_7g5; short heat shock protein 60 Hsp60s1; SPG13; Unknown (protein for IMAGE:7943409) | |
Recombinant Chlamydia trachomatis HSP60. | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.