Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP65 (mycobacterial) Antibody (3F7), Novus Biologicals™
SDP

Catalog No. NBP19788002 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19788002 0.2 mL
NBP19788005 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP19788002 Supplier Novus Biologicals Supplier No. NBP1978800.2ML
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HSP65 (mycobacterial) Monoclonal specifically detects HSP65 (mycobacterial) in Human, Mouse, Rat, Bacteria, Bovine, Drosophila, Guinea Pig, Hamster, Protozoa samples. It is validated for Western Blot.

Specifications

Antigen HSP65 (mycobacterial)
Applications Western Blot
Classification Monoclonal
Clone 3F7
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS with 0.09% Sodium Azide
Gene Alias 60 kDa chaperonin 2, 65 kDa antigen, Antigen A, Cell wall protein A, GroEL protein 2, groEL-2, groL2, Heat shock protein 65, hsp65, MT0456, MTV037.04, Protein Cpn60-2, Rv0440
Host Species Mouse
Immunogen Recombinant M. bovis HSP65.
Purification Method Ammonium sulfate precipitation
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 886354
Test Specificity Bacteria (E. coli, M. leprae, M. bovis), cryptosporidium
Target Species Human, Mouse, Rat, Bacteria, Bovine, Drosophila, Guinea Pig, Hamster, Protozoa
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.