Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSP70/HSC70 Monoclonal Antibody (BB70), PerCP
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA545101
This item is not returnable.
View return policy
Description
1 μg/mL of MA5-45101 was sufficient for detection of HSP70 and HSC70 in 20 μg of heat shocked HeLa cell lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 72 (HSP) and approximately 73kDa (HSC).
HSPA4 (Heat Shock Protein Family A (Hsp70) Member 4) is a Protein Coding gene. Diseases associated with HSPA4 include Chagas Disease and Babesiosis. Among its related pathways are Regulation of activated PAK-2p34 by proteasome mediated degradation and Cellular responses to stimuli. An important paralog of this gene is HSPH1. HSPA8 (Heat Shock Protein Family A (Hsp70) Member 8) is a Protein Coding gene. Diseases associated with HSPA8 include Auditory System Disease and Borna Disease. Among its related pathways are Processing of Capped Intron-Containing Pre-mRNA and Selective autophagy. Gene Ontology (GO) annotations related to this gene include RNA binding and ubiquitin protein ligase binding. An important paralog of this gene is HSPA2.
Specifications
HSP70/HSC70 | |
Monoclonal | |
1 mg/mL | |
2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.2 | |
O73885, O88600, P02827, P11142, P19120, P19378, P34932, P63017, P63018, Q2TFN9, Q61316, Q7YQC6 | |
hsp70, hspa1l.S, HSPA4, HSPA8 | |
Chicken HSP70/HSP90 complex. | |
200 μg | |
Primary | |
Human, Mouse, Rat, Canine, Rabbit, Bovine, Sheep, Hamster, Pig, Chicken, Fish, Drosophila, Yeast, Guinea Pig, Frog, Mammalia | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot, Immunocytochemistry | |
BB70 | |
PerCP | |
HSPA8 | |
HSPA4/HSPA8 | |
Mouse | |
Protein G | |
RUO | |
100037899, 100338049, 100348873, 100511890, 100524853, 100689472, 100713849, 100735343, 100764628, 101112844, 101122782, 15481, 15525, 24468, 266759, 281828, 281831, 3308, 3312, 379760, 395853, 403612, 416339, 474680, 479406, 536558 | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction