Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSP70/HSC70 Monoclonal Antibody (N27F3-4), APC
GREENER_CHOICE

Catalog No. PIMA545093
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545093 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545093 Supplier Invitrogen™ Supplier No. MA545093
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45093 was sufficient for detection of HSP70/HSC70 in 20 μg of heat shocked HeLa cell lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 72 (HSP) and approximately 73kDa (HSC).

HSPA4 (Heat Shock Protein Family A (Hsp70) Member 4) is a Protein Coding gene. Diseases associated with HSPA4 include Chagas Disease and Babesiosis. Among its related pathways are Regulation of activated PAK-2p34 by proteasome mediated degradation and Cellular responses to stimuli. An important paralog of this gene is HSPH1. HSPA8 (Heat Shock Protein Family A (Hsp70) Member 8) is a Protein Coding gene. Diseases associated with HSPA8 include Auditory System Disease and Borna Disease. Among its related pathways are Processing of Capped Intron-Containing Pre-mRNA and Selective autophagy. Gene Ontology (GO) annotations related to this gene include RNA binding and ubiquitin protein ligase binding. An important paralog of this gene is HSPA2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP70/HSC70
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N27F3-4
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.2
Gene HSPA4
Gene Accession No. O73885, O88600, P02827, P11142, P19120, P19378, P34932, P63017, P63018, Q2TFN9, Q61316, Q7YQC6
Gene Alias HSPA4/HSPA8
Gene Symbols hsp70, hspa1l.S, HSPA4, HSPA8
Host Species Mouse
Immunogen Recombinant HSP70/HSC70.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100037899, 100338049, 100348873, 100511890, 100524853, 100689472, 100713849, 100735343, 100764628, 101112844, 101122782, 15481, 15525, 24468, 266759, 281828, 281831, 3308, 3312, 379760, 395853, 403612, 416339, 474680, 479406, 536558
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Sheep, Hamster, Pig, Chicken, Chicken, Drosophila, Guinea Pig, Frog, C. elegans, Rat, X. laevis
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.