Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP70/HSPA1A Antibody (1H11), Novus Biologicals™

Mouse Monoclonal Antibody
$622.00
Specifications
Antigen | HSP70/HSPA1A |
---|---|
Clone | 1H11 |
Concentration | 0.5 mg/ml |
Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
Classification | Monoclonal |
Description
HSP70/HSPA1A Monoclonal specifically detects HSP70/HSPA1A in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface).Specifications
HSP70/HSPA1A | |
0.5 mg/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Pig, Bovine | |
3303 | |
Human native HSP70 protein | |
Primary | |
Binds the altered conformation of HSP70 embedded in the cell membrane and to intracellular HSP70 to a lesser extent. |
1H11 | |
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
Unconjugated | |
Mouse | |
Cancer, Cellular Markers, Chaperone Mediated Autophagy (CMA), Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific | |
dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B | |
HSPA1A | |
IgG1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title