Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP70/HSPA1A, p Ser153 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP260773
Description
HSP70/HSPA1A Polyclonal specifically detects HSP70/HSPA1A in Human samples. It is validated for Western Blot.Specifications
| HSP70/HSPA1A (p Ser153) | |
| Polyclonal | |
| Western Blot 1:100-1:2000 | |
| dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B | |
| Rabbit | |
| 70 kDa | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol. with No Preservative | |
| HSPA1A | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Thr153 conjugated to KLH. Accession # P0DMV8 | |
| Antigen Affinity-purified | |
| Cancer, Cellular Markers, Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific | |
| 3303 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction